missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethyl-1,3-dioxolane, 99%

Product Code. 11454857
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454857 500 g 500g
11434857 25 g 25g
11444857 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11454857 Supplier Thermo Scientific Alfa Aesar Supplier No. B21823.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1193-11-9 | C6H12O2 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1193-11-9
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00090841
InChI Key ALTFLAPROMVXNX-UHFFFAOYNA-N
PubChem CID 62384
IUPAC Name 2,2,4-trimethyl-1,3-dioxolane
SMILES CC1COC(C)(C)O1

Specifications

Density 0.897
Boiling Point 98°C
Flash Point 32°C (90°F)
Quantity 500 g
UN Number UN3271
Synonym propylene glycol acetone ketal, 1,3-dioxolane, 2,2,4-trimethyl, acetone propylene glycol acetal, 1,3-dioxolane,2,2,4-trimethyl, fema no. 3441, 2,2,4-trimethyl-1,3-oxacyclopentane, 2,2,4-trimethyl-1,3-dioxacyclopentane, propylene glycol acetonide, 1,2-propanediol acetonide, 2,2,4-trimethyldioxolane
Formula Weight 116.16
Percent Purity 99%
Chemical Name or Material 2,2,4-Trimethyl-1,3-dioxolane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.