missing translation for 'onlineSavingsMsg'
Learn More

2,2',4'-Trichloroacetophenone, 97%

Product Code. 11479792
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479792 100 g 100g
11469792 25 g 25g
11489792 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479792 Supplier Thermo Scientific Alfa Aesar Supplier No. A10835.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4252-78-2 | C8H5Cl3O | 223.477 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4252-78-2
Molecular Formula C8H5Cl3O
Molecular Weight (g/mol) 223.477
MDL Number MFCD00000934
InChI Key VYWPPRLJNVHPEU-UHFFFAOYSA-N
Synonym 2,2',4'-trichloroacetophenone, 2-chloro-1-2,4-dichlorophenyl ethanone, 2,4-dichlorophenacyl chloride, unii-2e35p7hji2, 2-chloro-1-2,4-dichlorophenyl ethan-1-one, ccris 633, ethanone, 2-chloro-1-2,4-dichlorophenyl, 2,2,4-trichloroacetophenone, dsstox_cid_6190, 1-2,4-dichlorophenyl-2-chloroethan-1-one
PubChem CID 20250
IUPAC Name 2-chloro-1-(2,4-dichlorophenyl)ethanone
SMILES C1=CC(=C(C=C1Cl)Cl)C(=O)CCl

Specifications

Melting Point 52°C to 56°C
Flash Point >110°C (230°F)
Quantity 100 g
UN Number UN2923
Beilstein 957098
Formula Weight 223.49
Percent Purity 97%
Chemical Name or Material 2,2',4'-Trichloroacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.