missing translation for 'onlineSavingsMsg'
Learn More

2-(2,4-Dihydroxybenzoyl)benzoic Acid, Mikromol™

Product Code. 30459373
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30459373 4 x 25 mg -
17923146 25 mg 25mg
17971346 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30459373 Supplier Mikromol Supplier No. MM0133.0400254AMP
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-(2,4-Dihydroxybenzoyl)benzoic Acid, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(2,4-Dihydroxybenzoyl)benzoic Acid
CAS 2513-33-9
Analyte or Component Names 2-(2,4-Dihydroxybenzoyl)benzoic Acid
Shipping Condition Room Temperature
Molecular Formula C14 H10 O5
Certifications/Compliance ISO 17025
Synonym Fluorescein Sodium Imp. C (EP), Fluorescein Imp. C (EP), Fluorescein USP Related Compound C, Fluorescein USP RC C, 2-(2,4-Dihydroxybenzoyl)benzoic Acid, Fluorescein Related Compound C, Fluorescein Sodium Impurity C, Fluorescein Impurity C
InChI Formula InChI=1S/C14H10O5/c15-8-5-6-11(12(16)7-8)13(17)9-3-1-2-4-10(9)14(18)19/h1-7,15-16H,(H,18,19)
Solution Type Neat
SMILES OC(=O)c1ccccc1C(=O)c2ccc(O)cc2O
IUPAC Name 2-(2,4-dihydroxybenzoyl)benzoic acid
Molecular Weight (g/mol) 258.23
Quantity 4 x 25 mg
Formula Weight 258.0528 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.