missing translation for 'onlineSavingsMsg'
Learn More

2-(2,3-dichlorophenyl)-1,3-thiazole-4-carboxylic acid, Thermo Scientific™

Product Code. 10557563
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10557563 1 g 1g
10382813 250 mg 250mg
10653635 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10557563 Supplier Thermo Scientific Maybridge Supplier No. SPB02181DA
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 257876-07-6
Molecular Formula C10H5Cl2NO2S
Molecular Weight (g/mol) 274.115
InChI Key ANTJCNIMRNBFMJ-UHFFFAOYSA-N
Synonym 2-2,3-dichlorophenyl-1,3-thiazole-4-carboxylic acid, 2-2,3-dichloro-phenyl-thiazole-4-carboxylic acid, 2-2,3-dichlorophenyl thiazole-4-carboxylic acid, 4-thiazolecarboxylicacid, 2-2,3-dichlorophenyl
PubChem CID 2743987
IUPAC Name 2-(2,3-dichlorophenyl)-1,3-thiazole-4-carboxylic acid
SMILES C1=CC(=C(C(=C1)Cl)Cl)C2=NC(=CS2)C(=O)O

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 274.13
Percent Purity 97%
Chemical Name or Material 2-(2, 3-Dichlorophenyl)-1, 3-thiazole-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.