Product Code. 30136725
Click to view available options
Quantity:
1 mL
2.5 mL
5 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
unitSize
30136725
2.5 mL
2.5mL
30011098
5 mL
5mL
30004141
1 mL
1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code.
30136725
Supplier
Toronto Research Chemicals
Supplier No.
TRCP2272252.5ML
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Chemical Identifiers
CAS
13475-82-6
Molecular Formula
C12 H26
Molecular Weight (g/mol)
170.33
Synonym
Sodium saccharin, Sodium 2-benzimidazole-1,1-dioxide
IUPAC Name
2,2,4,6,6-pentamethylheptane
SMILES
CC(CC(C)(C)C)CC(C)(C)C
Specifications
Quantity
2.5 mL
InChI Formula
InChI=1S/C12H26/c1-10(8-11(2,3)4)9-12(5,6)7/h10H,8-9H2,1-7H3
Formula Weight
170.2035
Chemical Name or Material
2,2,4,6,6-Pentamethylheptane
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?