missing translation for 'onlineSavingsMsg'
Learn More

2,2,4,6,6-Pentamethylheptane

Product Code. 16544214 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16544214 5 mL 5mL
16554214 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 16544214 Supplier Thermo Scientific Acros Supplier No. 462050050
  • ONLINE EXCLUSIVE PRICE
    £50.10
    £45.65 / 5mL
    Save £4.45
    8% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 13475-82-6 | C12H26 | 170.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13475-82-6
Molecular Formula C12H26
Molecular Weight (g/mol) 170.34
InChI Key VKPSKYDESGTTFR-UHFFFAOYSA-N
IUPAC Name 2,2,4,6,6-pentamethylheptane
SMILES CC(CC(C)(C)C)CC(C)(C)C

Specifications

Density 0.7463 g/mL
Boiling Point 170.0°C
Flash Point 37°C
Linear Formula [CH3C(CH3)2CH2]2CHCH3
Quantity 5 mL
Specific Gravity 0.7463
Formula Weight 170.33
Chemical Name or Material 2,2,4,6,6-Pentamethylheptane
compliance-icons compliance-icons
Product Identifier
  • 2,2,4,6,6-Pentamethylheptane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 4
  • Aspiration toxicity Category 1
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H413-May cause long lasting harmful effects to aquatic life.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P331-Do NOT induce vomiting.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contain: 2,2,4,6,6-Pentamethylheptane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.