missing translation for 'onlineSavingsMsg'
Learn More

2-(2,4,5-Trichlorophenoxy)propionic acid, 96%, Thermo Scientific™

Product Code. 11308005
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11308005 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11308005 Supplier Thermo Scientific Alfa Aesar Supplier No. L08136.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-72-1
Molecular Formula C9H7Cl3O3
Molecular Weight (g/mol) 269.50
MDL Number MFCD00002646
InChI Key ZLSWBLPERHFHIS-UHFFFAOYNA-N
Synonym fenoprop, silvex, 2-2,4,5-trichlorophenoxy propionic acid, double strength, fenormone, propon, color-set, aqua-vex, sta-fast, fruitone t
PubChem CID 7158
ChEBI CHEBI:34758
IUPAC Name 2-(2,4,5-trichlorophenoxy)propanoic acid
SMILES CC(OC1=CC(Cl)=C(Cl)C=C1Cl)C(O)=O

Specifications

Melting Point 176°C to 180°C
Quantity 25 g
UN Number UN3077
Beilstein 1985768
Formula Weight 269.51
Percent Purity 96%
Chemical Name or Material 2-(2,4,5-Trichlorophenoxy)propionic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.