missing translation for 'onlineSavingsMsg'
Learn More

2,2',4,4'-Tetrahydroxybenzophenone, TRC

Product Code. 17982665
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17982665 5 g 5g
17913725 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17982665 Supplier Toronto Research Chemicals Supplier No. TRCT2959005G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131-55-5
Molecular Formula C13 H10 O5
Molecular Weight (g/mol) 246.2155
Synonym Benzophenone, 2,2',4,4'-tetrahydroxy- (6CI,8CI), Bis(2,4-dihydroxyphenyl)methanone, 2,2',4,4'-Tetrahydroxy diphenyl ketone, 2,2',4,4'-Tetrahydroxybenzophenone, 2,4,2',4'-Tetrahydroxybenzophenone, BP 2, Benzophenone 2, Dainsorb P 6, Eversorb 51, NSC 38556, SUV 1, Seesorb 106, Sumisorb 150, T 0118, Uvinul 3050, Uvinul D 50
IUPAC Name bis(2,4-dihydroxyphenyl)methanone
SMILES Oc1ccc(C(=O)c2ccc(O)cc2O)c(O)c1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C13H10O5/c14-7-1-3-9(11(16)5-7)13(18)10-4-2-8(15)6-12(10)17/h1-6,14-17H
Formula Weight 246.0528
Percent Purity >95%
Chemical Name or Material 2,2',4,4'-Tetrahydroxybenzophenone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.