missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,4,4,6,8,8-Heptamethylnonane, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Color | Colorless |
Density | 0.7900g/mL |
Boiling Point | 240.0°C |
Flash Point | 95°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass Bottle |
Linear Formula | (CH3)3CCH2CH(CH3)CH2C(CH3)2CH2C(CH3)3 |
Show More |
Safety and Handling
Product Identifier
- 2,2,4,4,6,8,8-Heptamethylnonane
- Danger
- Aspiration toxicity Category 1
- Acute toxicity Category 4
- Specific target organ toxicity Category 2
- H302-Harmful if swallowed.
- H304-May be fatal if swallowed and enters airways.
- H332-Harmful if inhaled.
- H371-May cause damage to organs.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'