missing translation for 'onlineSavingsMsg'
Learn More

2,2,4,4,6,8,8-Heptamethylnonane, 98%, Thermo Scientific Chemicals

Catalog Number 10275960
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
4390-04-9
C16H34
226.44
MFCD00008856
VCLJODPNBNEBKW-UHFFFAOYNA-N
isocetane, nonane, 2,2,4,4,6,8,8-heptamethyl, cyprane, pubchem16042, acmc-2097fb, dsstox_cid_30668, dsstox_gsid_52101, ksc492g9j, 2,4,4,6,8,8-heptamethylnonane, nonane,2,4,4,6,8,8-heptamethyl
20414
2,2,4,4,6,8,8-heptamethylnonane
CC(CC(C)(C)C)CC(C)(C)CC(C)(C)C
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
4390-04-9
C16H34
226.44
MFCD00008856
VCLJODPNBNEBKW-UHFFFAOYNA-N
isocetane, nonane, 2,2,4,4,6,8,8-heptamethyl, cyprane, pubchem16042, acmc-2097fb, dsstox_cid_30668, dsstox_gsid_52101, ksc492g9j, 2,4,4,6,8,8-heptamethylnonane, nonane,2,4,4,6,8,8-heptamethyl
20414
2,2,4,4,6,8,8-heptamethylnonane
CC(CC(C)(C)C)CC(C)(C)CC(C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.7900g/mL
Boiling Point 240.0°C
Flash Point 95°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CCH2CH(CH3)CH2C(CH3)2CH2C(CH3)3
Refractive Index 1.4380 to 1.4400
Quantity 50 mL
Specific Gravity 0.79
Solubility Information Solubility in water: insoluble
Formula Weight 226.44
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2,2,4,4,6,8,8-Heptamethylnonane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,2,4,4,6,8,8-Heptamethylnonane
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H304-May be fatal if swallowed and enters airways.
  • H332-Harmful if inhaled.
  • H371-May cause damage to organs.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'