missing translation for 'onlineSavingsMsg'
Learn More

2,2,4,4,6,8,8-Heptamethylnonane, 98.3%, MP Biomedicals™

Product Code. 11483802 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11483802 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11483802 Supplier MP Biomedicals Supplier No. 0215732280

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 4393-09-3
Color Colorless
Assay Percent Range 98.3%
Molecular Formula C9H13NO2
Quantity 100 mL
Synonym 2,3-dimethoxybenzylamine, 2,3-dimethoxyphenyl methanamine, 1-2,3-dimethoxyphenyl methanamine, benzenemethanamine, 2,3-dimethoxy, 2,3-dimethoxy-benzylamine, 2,3-dimethoxy benzylamine, pubchem7399, 2,3-dimethoxybenzyl amine, acmc-2097fc
InChI Key LVMPWFJVYMXSNY-UHFFFAOYSA-N
SMILES COC1=CC=CC(=C1OC)CN
IUPAC Name (2,3-dimethoxyphenyl)methanamine
Molecular Weight (g/mol) 167.21
PubChem CID 78106
Formula Weight 226.45
Physical Form Liquid
Chemical Name or Material 2,2,4,4,6,8,8-Heptamethylnonane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.