missing translation for 'onlineSavingsMsg'
Learn More

2’-(2,3-Epoxypropoxy)-3-phenylpropiophenone, TRC

Product Code. 30380190
Change view
Click to view available options
Quantity:
25 mg
250 mg
5 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380190 25 mg -
30384009 5 g 5g
30383263 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380190 Supplier Toronto Research Chemicals Supplier No. TRCE58970025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22525-95-7
Molecular Formula C18 H18 O3
Molecular Weight (g/mol) 282.33
Synonym 1-[2-[[(2RS)-Oxiranyl]methoxy]phenyl]-3-phenylpropan-1-one, Propafenone Hydrochloride Imp. C (EP), 1-Propanone, 1-[2-(oxiranylmethoxy)phenyl]-3-phenyl- (9CI), Propiophenone, 2'-(2,3-epoxypropoxy)-3-phenyl- (8CI), 2-[[2-(3-Phenylpropanoyl)phenoxy]methyl]oxirane, 2'-(2,3-Epoxypropoxy)-3-phenylpropiophenone
IUPAC Name 1-[2-(oxiran-2-ylmethoxy)phenyl]-3-phenylpropan-1-one
SMILES O=C(CCc1ccccc1)c2ccccc2OCC3CO3

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C18H18O3/c19-17(11-10-14-6-2-1-3-7-14)16-8-4-5-9-18(16)21-13-15-12-20-15/h1-9,15H,10-13H2
Formula Weight 282.1256
Chemical Name or Material 1-[2-[((2RS)-Oxiranyl)methoxy]phenyl]-3-phenylpropan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.