missing translation for 'onlineSavingsMsg'
Learn More

2-(2,3-Dihydro-1-benzofuran-5-yl)-4-methyl-1,3-thiazole-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10255904
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10255904 5 g 5g
10291133 250 mg 250mg
10666323 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10255904 Supplier Thermo Scientific Maybridge Supplier No. SP00239DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 690632-04-3
Molecular Formula C13H11NO3S
Molecular Weight (g/mol) 261.295
MDL Number MFCD01313483
InChI Key DCXMMNGTUXDRND-UHFFFAOYSA-N
Synonym 2-2,3-dihydro-1-benzofuran-5-yl-4-methyl-1,3-thiazole-5-carboxylic acid, 2-2,3-dihydrobenzofuran-5-yl-4-methylthiazole-5-carboxylic acid, 2-2,3-dihydro-1-benzo b furan-5-yl-4-methyl-1,3-thiazole-5-carboxylic acid, 2-2,3-dihydrobenzo b furan-5-yl-4-methyl-1,3-thiazole-5-carboxylic acid
PubChem CID 2798545
IUPAC Name 2-(2,3-dihydro-1-benzofuran-5-yl)-4-methyl-1,3-thiazole-5-carboxylic acid
SMILES CC1=C(SC(=N1)C2=CC3=C(C=C2)OCC3)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 261.3
Percent Purity 97%
Chemical Name or Material 2-(2, 3-Dihydro-1-benzofuran-5-yl)-4-methyl-1, 3-thiazole-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.