missing translation for 'onlineSavingsMsg'
Learn More

2-(2,3-Dihydro-1-benzofuran-5-yl)-1,3-thiazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 11376945
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
Product Code. Quantity Packaging unitSize
11376945 5 g Amber Glass Bottle 5g
10713843 1 g Amber Glass Bottle 1g
2 options
This item is not returnable. View return policy

Product Code. 11376945

Brand: Thermo Scientific Maybridge SP00242DE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 368869-97-0
Molecular Formula C12H9NO3S
Molecular Weight (g/mol) 247.27
MDL Number MFCD01313485
InChI Key NOUOFPSRFBTJCH-UHFFFAOYSA-N
Synonym 2-2,3-dihydro-1-benzofuran-5-yl-1,3-thiazole-4-carboxylic acid, 2-2,3-dihydrobenzofuran-5-yl thiazole-4-carboxylic acid, 4-thiazolecarboxylicacid, 2-2,3-dihydro-5-benzofuranyl, maybridge1_008369, 2-2,3-dihydrobenzofuran-5-yl-4-thiazolecarboxylic acid, 2-2,3-dihydrobenzo b furan-5-yl-1,3-thiazole-4-carboxylic acid, 2-2,3-dihydro-1-benzo b furan-5-yl-1,3-thiazole-4-carboxylic acid
PubChem CID 2742272
IUPAC Name 2-(2,3-dihydro-1-benzofuran-5-yl)-1,3-thiazole-4-carboxylic acid
SMILES OC(=O)C1=CSC(=N1)C1=CC2=C(OCC2)C=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 247.27
Percent Purity 97%
Chemical Name or Material 2-(2, 3-Dihydro-1-benzofuran-5-yl)-1, 3-thiazole-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.