missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2',3,4,4',5'-Hexabromodiphenyl Ether, TRC
Specifications
Specifications
| Chemical Name or Material | PBDE 138 (2,2',3,4,4',5'-Hexabromodiphenyl Ether) |
| CAS | 182677-30-1 |
| Molecular Formula | C12 H4 Br6 O |
| Quantity | 500 mg |
| Synonym | PBDE No. 138, Benzene, 1,2,3-tribromo-4-(2,4,5-tribromophenoxy)-, 1,2,3-Tribromo-4-(2,4,5-tribromophenoxy)benzene, 2,2',3,4,4',5'-Hexabromodiphenyl ether, BDE 138, PBDE 138, PBDE 138 (2,2',3,4,4',5'-Hexabromodiphenyl Ether), Benzene, 1,2,3-tribromo-4-(2,4,5-tribromophenoxy)-, 1,2,3-Tribromo-4-(2,4,5-tribromophenoxy)benzene, 2,2',3,4,4',5'-Hexabromodiphenyl ether, 2,2',3,4,4',5'-HexaBDE, BDE 138, PBDE 138 |
| InChI Formula | InChI=1S/C12H4Br6O/c13-5-1-2-9(12(18)11(5)17)19-10-4-7(15)6(14)3-8(10)16/h1-4H |
| SMILES | Brc1cc(Br)c(Oc2ccc(Br)c(Br)c2Br)cc1Br |
| IUPAC Name | 1,2,3-tribromo-4-(2,4,5-tribromophenoxy)benzene |
| Molecular Weight (g/mol) | 643.58 |
| Formula Weight | 637.5362 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?