missing translation for 'onlineSavingsMsg'
Learn More

2,2',3,4,4',5,5'-Heptachlorobiphenyl, TRC

Product Code. 30389071
Change view
Click to view available options
Quantity:
10 mg
25 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30389071 250 mg
30396425 25 mg
30390996 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30389071 Supplier Toronto Research Chemicals Supplier No. TRCH265060250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35065-29-3
Molecular Formula C12 H3 Cl7
Molecular Weight (g/mol) 395.32
Synonym 1,1'-Biphenyl, 2,2',3,4,4',5,5'-heptachloro-, 2,2',3,4,4',5,5'-Heptachloro-1,1'-biphenyl, 2,2',3,4,4',5,5'-Heptachlorobiphenyl, 2,3,4,5,2',4',5'-Heptachlorobiphenyl, CB 180, K 180, PCB 180, 2,2',3,4,4',5,5'-HeptaCB
IUPAC Name 1,2,3,4-tetrachloro-5-(2,4,5-trichlorophenyl)benzene
SMILES Clc1cc(Cl)c(cc1Cl)c2cc(Cl)c(Cl)c(Cl)c2Cl

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C12H3Cl7/c13-6-3-8(15)7(14)1-4(6)5-2-9(16)11(18)12(19)10(5)17/h1-3H
Formula Weight 391.8054
Percent Purity >95%
Chemical Name or Material PCB No. 180
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.