missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,4,4,4-Hexafluorobutyl methacrylate, 96%, stab.

Product Code. 11307486
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11307486 25 g 25g
11397476 5 g 5g
11317486 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11307486 Supplier Thermo Scientific Alfa Aesar Supplier No. L09543.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 36405-47-7 | C8H8F6O2 | 250.14 g/mol

It is an epoxy thinner and is mainly used as the modifier of the (meth)acrylate resin. Well-defined organic/inorganic hybrid fluorinated star polymers were synthesized via atom transfer radical polymerization (ATRP) of 2,2,3,4,4,4-hexafluorobutyl methacrylate (HFBMA) using octa(aminophenyl)silsesquioxane (OAPS) nano-cage as initiator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an epoxy thinner and is mainly used as the modifier of the (meth)acrylate resin. Well-defined organic/inorganic hybrid fluorinated star polymers were synthesized via atom transfer radical polymerization (ATRP) of 2,2,3,4,4,4-hexafluorobutyl methacrylate (HFBMA) using octa(aminophenyl)silsesquioxane (OAPS) nano-cage as initiator.

Solubility
Difficult to mix in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36405-47-7
Molecular Formula C8H8F6O2
Molecular Weight (g/mol) 250.14
MDL Number MFCD00042311
InChI Key DFVPUWGVOPDJTC-UHFFFAOYSA-N
Synonym 2,2,3,4,4,4-hexafluorobutyl methacrylate, 1h,1h,3h-hexafluorobutyl methacrylate, hexafluorobutyl methacrylate, methacrylic acid 2,2,3,4,4,4-hexafluorobutyl ester, 2-propenoic acid, 2-methyl-, 2,2,3,4,4,4-hexafluorobutyl ester, hexafluorobutyl methacrylate polymer, hfbma, acmc-1adv6, ksc489s1j, 2-propenoic acid, 2-methyl-, 2,2,3,4,4,4-hexafluorobutyl ester, homopolymer
PubChem CID 549772
IUPAC Name 2,2,3,4,4,4-hexafluorobutyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC(C(C(F)(F)F)F)(F)F

Specifications

Density 1.352
Boiling Point 158°C
Flash Point 56°C (132°F)
Refractive Index 1.362
Quantity 25 g
UN Number UN3272
Beilstein 2725177
Solubility Information Difficult to mix in water.
Formula Weight 250.14
Percent Purity 96%
Chemical Name or Material 2,2,3,4,4,4-Hexafluorobutyl methacrylate, stabilized
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.