missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,3,3-Tetramethylsuccinonitrile, TRC
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
Specifications
Specifications
| Chemical Name or Material | Tetramethylsuccinonitrile |
| CAS | 3333-52-6 |
| Molecular Formula | C8 H12 N2 |
| Quantity | 5 g |
| Synonym | 2,2,3,3-Tetramethylbutanedinitrile, Butanedinitrile','tetramethyl- (9CI), Succinonitrile','tetramethyl- (6CI,7CI,8CI), 2,2,3,3-Tetramethylbutanedinitrile, 2,2,3,3-Tetramethylsuccinonitrile, 2,3-Dicyano-2,3-dimethylbutane, NSC 39746, Tetramethylsuccinic acid |
| InChI Formula | InChI=1S/C8H12N2/c1-7(2,5-9)8(3,4)6-10/h1-4H3 |
| SMILES | CC(C)(C#N)C(C)(C)C#N |
| IUPAC Name | 2,2,3,3-tetramethylbutanedinitrile |
| Molecular Weight (g/mol) | 136.194 |
| Formula Weight | 136.1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction