missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3-Tetramethylcyclopropanecarboxylic acid, 97%, Thermo Scientific™

Product Code. 15476026
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15476026 1 g 1g
15486026 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15476026 Supplier Thermo Scientific Alfa Aesar Supplier No. H60809.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2,2,3,3-Tetramethylcyclopropanecarboxylic acid is used to start synthesis of central nervous system-active compounds.

Solubility
Slightly soluble in water. Soluble in methanol, ethanol and acetonitrile.

Notes
Keep container tightly sealed. Store in refrigerated conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15641-58-4
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.198
MDL Number MFCD00011538
InChI Key SFHVXKNMCGSLAR-UHFFFAOYSA-N
Synonym 2,2,3,3-tetramethylcyclopropanecarboxylic acid, tetramethylcyclopropane-1-carboxylic acid, cyclopropanecarboxylic acid, 2,2,3,3-tetramethyl, 2,2,3,3-tetramethylcyclopropanecarboxylicacid, acmc-1c48r, tetramethylcyclopropanecarboxylicacid, 3-carboxy-1,1,2,2-tetramethylcyclopropane, 2,2,3,3-tetramethyl cyclopropanecarboxylic acid, 2,2,3,3-tetramethyl-cyclopropanecarboxylic acid, 2,2,3,3-tetramethylcyclopropane carboxylic acid
PubChem CID 152115
ChEBI CHEBI:39355
IUPAC Name 2,2,3,3-tetramethylcyclopropane-1-carboxylic acid
SMILES CC1(C(C1(C)C)C(=O)O)C

Specifications

Melting Point 120°C to 121°C
Quantity 1 g
Solubility Information Slightly soluble in water. Soluble in methanol,ethanol and acetonitrile.
Formula Weight 142.2
Percent Purity 97%
Chemical Name or Material 2,2,3,3-Tetramethylcyclopropanecarboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.