missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3-Tetrafluoro-1,4-butanediol, 97%

Product Code. 11349295
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11349295 25 g 25g
11329295 1 g 1g
11339295 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11349295 Supplier Thermo Scientific Alfa Aesar Supplier No. L16635.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 425-61-6 | C4H6F4O2 | 162.08 g/mol

2,2,3,3-Tetrafluoro-1,4-butanediol is a reactant used for the enzymic synthesis of silicone fluorinated aliphatic polyester amides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,3,3-Tetrafluoro-1,4-butanediol is a reactant used for the enzymic synthesis of silicone fluorinated aliphatic polyester amides.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with acids and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 425-61-6
Molecular Formula C4H6F4O2
Molecular Weight (g/mol) 162.08
MDL Number MFCD00042375
InChI Key CDZXJJOGDCLNKX-UHFFFAOYSA-N
Synonym 2,2,3,3-tetrafluoro-1,4-butanediol, 1,4-dihydroxy-2,2,3,3-tetrafluorobutane, acmc-1aqx7, 1,4-butanediol,2,2,3,3-tetrafluoro, 2,2,3,3-tetrakis fluoranyl butane-1,4-diol
PubChem CID 136270
IUPAC Name 2,2,3,3-tetrafluorobutane-1,4-diol
SMILES OCC(F)(F)C(F)(F)CO

Specifications

Melting Point 80°C to 87°C
Boiling Point 110°C to 112°C (13 mmHg)
Quantity 25 g
Beilstein 1745847
Solubility Information Soluble in water.
Formula Weight 162.08
Percent Purity 97%
Chemical Name or Material 2,2,3,3-Tetrafluoro-1,4-butanediol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.