missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3-d(4)-3-(Trimethylsilyl)propionic acid sodium salt, 98+ atom % D

Product Code. 11434130
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11434130 250 mg 250mg
11471818 1 g 1g
11481818 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11434130 Supplier Thermo Scientific Alfa Aesar Supplier No. A14489.MD
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 24493-21-8 | C6H13NaO2Si | 172.267 g/mol

2,2,3,3-d(4)-3-(Trimethylsilyl)propionic acid sodium salt is used as a deuterium labeled silane. Also used in pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,3,3-d(4)-3-(Trimethylsilyl)propionic acid sodium salt is used as a deuterium labeled silane. Also used in pharmaceutical industry.

Solubility
Soluble in water.

Notes
It is hygroscopic. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24493-21-8
Molecular Formula C6H13NaO2Si
Molecular Weight (g/mol) 172.267
MDL Number MFCD00002762
InChI Key OIIWPAYIXDCDNL-HGFPCDIYSA-M
Synonym sodium 3-trimethylsilyl 2,2,3,3-2h4 propionate, 3-trimethylsilyl propionic acid-d4 sodium salt, 2,2,3,3-d4-3-trimethylsilyl propionic acid sodium salt, sodium 3-trimethylsilyl 2 h? propanoate, sodium 3-trimethylsilyl 2h4 propanoate, tsp-d4, propanoic-2,2,3,3-d4 acid, 3-trimethylsilyl-, sodium salt, sodium 3-trimethylsilyl propionate-2,2,3,3-d4, 3-trimethylsilyl propionic-2,2,3,3-d4 acid sodium salt, sodium-3-trimethylsilyl-2,2,3,3-tetradeuteriopropionate
PubChem CID 23688921
IUPAC Name sodium;2,2,3,3-tetradeuterio-3-trimethylsilylpropanoate
SMILES C[Si](C)(C)CCC(=O)[O-].[Na+]

Specifications

Melting Point >300°C
Linear Formula (CH3)3SiCD2CD2CO2Na
Quantity 250 mg
Beilstein 4046239
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 172.27
Assay ≥98 atom %D
Chemical Name or Material 2,2,3,3-d(4)-3-(Trimethylsilyl)propionic acid sodium salt
compliance-icons
Product Identifier
  • 2,2,3,3-d{ 4} -3-(Trimethylsilyl)propionicacid, sodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2,2,3,3-d{ 4} -3-(Trimethylsilyl)propionicacid, sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.