missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,4,4,4-Heptafluorobutylamine, 97%, Thermo Scientific Chemicals

Product Code. 11383736
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11383736 1 g 1g
11393736 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11383736 Supplier Thermo Scientific Alfa Aesar Supplier No. L16776.03
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 374-99-2 | Molecular Formula: C4H4F7N | Molecular Weight: 199.072 g/mol

2,2,3,3,4,4,4-Heptafluorobutylamine is used as pharmaceutical intermediates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,3,3,4,4,4-Heptafluorobutylamine is used as pharmaceutical intermediates

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 374-99-2
Molecular Formula C4H4F7N
Molecular Weight (g/mol) 199.072
MDL Number MFCD00014817
InChI Key WBGBQSRNXPVFDB-UHFFFAOYSA-N
Synonym 2,2,3,3,4,4,4-heptafluorobutylamine, 1h,1h-heptafluorobutylamine, heptafluorobutylamine, butylamine, 2,2,3,3,4,4,4-heptafluoro, 1-butanamine, 2,2,3,3,4,4,4-heptafluoro, 1h,1h-perfluorobutylamine, acmc-1af94, 1-butanamine,2,2,3,3,4,4,4-heptafluoro
PubChem CID 67807
IUPAC Name 2,2,3,3,4,4,4-heptafluorobutan-1-amine
SMILES C(C(C(C(F)(F)F)(F)F)(F)F)N

Specifications

Boiling Point 70°C to 71°C
Quantity 1 g
UN Number UN2735
Beilstein 1761906
Formula Weight 199.07
Percent Purity 97%
Chemical Name or Material 2,2,3,3,4,4,4-Heptafluorobutylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.