missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,4,4,4-Heptafluoro-1-iodobutane, 96%

Product Code. 11336826
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11336826 1 g 1g
11346826 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11336826 Supplier Thermo Scientific Alfa Aesar Supplier No. L04173.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 374-98-1 | C4H2F7I | 309.953 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 374-98-1
Molecular Formula C4H2F7I
Molecular Weight (g/mol) 309.953
MDL Number MFCD00013719
InChI Key AFHPVXVEXINDFS-UHFFFAOYSA-N
Synonym heptafluorobutyl iodide, 2,2,3,3,4,4,4-heptafluoro-1-iodobutane, butane, 1,1,1,2,2,3,3-heptafluoro-4-iodo, 1h,1h-heptafluorobutyl iodide, 1h,1h-heptafluoro-1-iodobutane, acmc-20aoko, 1-iodo-1h,1h-perfluorobutane, 1h,1h-heptafluoro-1-iodobutan, 1-iodo-2,2,3,3,4,4,4-heptafluorobutane
PubChem CID 67806
IUPAC Name 1,1,1,2,2,3,3-heptafluoro-4-iodobutane
SMILES C(C(C(C(F)(F)F)(F)F)(F)F)I

Specifications

Density 2.054
Boiling Point 91°C to 92°C
Flash Point None
Linear Formula CF3CF2CF2CH2I
Refractive Index 1.3625
Quantity 1 g
Beilstein 1761927
Sensitivity Light sensitive
Formula Weight 309.95
Percent Purity 96%
Chemical Name or Material 2,2,3,3,4,4,4-Heptafluoro-1-iodobutane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.