missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,4,4,4-Heptafluoro-1-butanol, 95%

Product Code. 15710849 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
Packaging:
Glass bottle
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15710849 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15710849 Supplier Thermo Scientific Acros Supplier No. 243890500
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 375-01-9 | C4H3F7O | 200.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 375-01-9
Molecular Formula C4H3F7O
Molecular Weight (g/mol) 200.06
MDL Number MFCD00004674
InChI Key WXJFKAZDSQLPBX-UHFFFAOYSA-N
Synonym 2,2,3,3,4,4,4-heptafluoro-1-butanol, 1-butanol, 2,2,3,3,4,4,4-heptafluoro, 1h,1h-heptafluorobutanol-1, 1h,1h-heptafluoro-1-butanol, 1,1-dihydroperfluorobutyl alcohol, 2,2,3,3,4,4,4-heptafluorobutanol, 1,1-dihydroperfluorobutanol, 1,1-h,h-heptafluorobutanol, perfluoro-1,1-dihydrobutanol, butanol, 2,2,3,3,4,4,4-heptafluoro
PubChem CID 9776
IUPAC Name 2,2,3,3,4,4,4-heptafluorobutan-1-ol
SMILES OCC(F)(F)C(F)(F)C(F)(F)F

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color White to Beige
Density 1.6000g/mL
Boiling Point 96°C to 97°C
Flash Point 25°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CF3CF2CF2CH2OH
Refractive Index 1.2920 to 1.294
Quantity 50 g
Beilstein 01, 1547
Specific Gravity 1.6
Solubility Information Solubility in water: slightly soluble
Formula Weight 200.06
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 2,2,3,3,4,4,4-Heptafluoro-1-butanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.