missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,3-Pentafluoropropanoic Acid, TRC

Product Code. 17923095
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17923095 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17923095 Supplier Toronto Research Chemicals Supplier No. TRCP2272351G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 422-64-0
Molecular Formula C3HF5O2
Quantity 1 g
Synonym Perfluoropropionic Acid, 2,2,3,3,3-Pentafluoropropanoic acid (ACI), Propanoic acid, pentafluoro- (9CI), Propionic acid, pentafluoro- (6CI, 7CI, 8CI), 2,2,3,3,3-Pentafluoropropionic acid, Pentafluoropropanoic acid, Pentafluoropropionic acid, Perfluoropropanoic acid, Perfluoropropionic acid, PFPrA, PFPrA, Pentafluoro-propanoic Acid, Pentafluoro-propionic Acid, 2,2,3,3,3-Pentafluoropropionic Acid, Pentafluoropropanoic Acid, Pentafluoropropionic Acid, Perfluoropropanoic Acid, Perfluoropropionic Acid
InChI Formula InChI=1S/C3HF5O2/c4-2(5,1(9)10)3(6,7)8/h(H,9,10)
SMILES OC(=O)C(F)(F)C(F)(F)F
IUPAC Name 2,2,3,3,3-pentafluoropropanoic acid
Molecular Weight (g/mol) 164.03
Formula Weight 163.9897
Chemical Name or Material 2,2,3,3,3-Pentafluoropropanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.