missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,3-Pentafluoropropanoic Acid-13C, TRC

Product Code. 30132791
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132791 100 mg 100mg
30010118 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30132791 Supplier Toronto Research Chemicals Supplier No. TRCP227237100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula 13C C2 H F5 O2
Molecular Weight (g/mol) 165.024
Synonym Pentafluoro-propanoic-13C Acid, Pentafluoro-propionic-13C Acid, 2,2,3,3,3-Pentafluoropropionic-13C Acid, Pentafluoropropanoic-13C Acid, Pentafluoropropionic-13C Acid, Perfluoropropanoic-13C Acid, Perfluoropropionic-13C Acid
IUPAC Name 2,2,3,3,3-pentafluoro(1^{13}C)propanoic acid
SMILES O[13C](=O)C(F)(F)C(F)(F)F

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C3HF5O2/c4-2(5,1(9)10)3(6,7)8/h(H,9,10)/i1+1
Formula Weight 164.993
Percent Purity >95%
Chemical Name or Material 2,2,3,3,3-Pentafluoropropanoic Acid-13C
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.