missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,3,3,3-Pentafluoro-1-propanol, 98%, Thermo Scientific Chemicals
81.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
2,2,3,3,3-Pentafluoro-1-propanol is used as Pharmaceutical Intermediates. It can be used in agrochemical, pharmaceutical and dyestuff field etc.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,2,3,3,3-Pentafluoro-1-propanol is used as Pharmaceutical Intermediates. It can be used in agrochemical, pharmaceutical and dyestuff field etc.
Solubility
Soluble in water.21.9 g/L at 25°C
Notes
Incompatible with strong oxidizing agents, Acid chlorides, Acid anhydrides. Keep away from heat.
2,2,3,3,3-Pentafluoro-1-propanol is used as Pharmaceutical Intermediates. It can be used in agrochemical, pharmaceutical and dyestuff field etc.
Solubility
Soluble in water.21.9 g/L at 25°C
Notes
Incompatible with strong oxidizing agents, Acid chlorides, Acid anhydrides. Keep away from heat.
Specifications
Density | 1.505 |
Boiling Point | 80°C to 81°C |
Refractive Index | 1.288 |
Quantity | 25 g |
Beilstein | 1743133 |
Solubility Information | Soluble in water.21.9g/L at 25°C |
Formula Weight | 150.05 |
Percent Purity | 98% |
Chemical Name or Material | 2,2,3,3,3-Pentafluoro-1-propanol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'