missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,3-Pentafluoro-1-propanol, 97%

Product Code. 10043650 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10043650 25 g Glass bottle 25g
10790581 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.
Product Code. 10043650 Supplier Thermo Scientific Acros Supplier No. 303790250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.

CAS: 422-05-9 | C3H3F5O | 150.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 422-05-9
Molecular Formula C3H3F5O
Molecular Weight (g/mol) 150.05
MDL Number MFCD00004673
InChI Key PSQZJKGXDGNDFP-UHFFFAOYSA-N
Synonym 2,2,3,3,3-pentafluoro-1-propanol, pentafluoro-1-propanol, 1,1-dihydroperfluoropropanol, pfpoh, 2,2,3,3,3-pentafluoropropanol, 1h,1h-pentafluoropropanol, 1-propanol, 2,2,3,3,3-pentafluoro, 1h,1h-pentafluoropropanol-1, c2f5ch2oh, unii-epj1s961sn
PubChem CID 9872
IUPAC Name 2,2,3,3,3-pentafluoropropan-1-ol
SMILES C(C(C(F)(F)F)(F)F)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color White to Yellow
Boiling Point 81.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CF3CF2CH2OH
Refractive Index 1.2880 to 1.2950
Quantity 25 g
Beilstein 01, IV, 1438
Formula Weight 150.05
Percent Purity 97%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material 2,2,3,3,3-Pentafluoro-1-propanol, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.