missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trimethylacetamide, 98+%

Product Code. 11463714
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463714 50 g 50g
11453714 10 g 10g
11473714 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11463714 Supplier Thermo Scientific Alfa Aesar Supplier No. A11319.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 754-10-9 | C5H11NO | 101.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 754-10-9
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.15
MDL Number MFCD00008011
InChI Key XIPFMBOWZXULIA-UHFFFAOYSA-N
PubChem CID 12957
IUPAC Name 2,2-dimethylpropanamide
SMILES CC(C)(C)C(N)=O

Specifications

Melting Point 153°C to 157°C
Boiling Point 212°C
Linear Formula (CH3)3CCONH2
Quantity 50 g
Beilstein 1742147
Synonym pivalamide, trimethylacetamide, 2,2,2-trimethylacetamide, propanamide, 2,2-dimethyl, 2,2-dimethylpropionamide, 2,2-dimethyl-propionamide, unii-fes86mr7zi, fes86mr7zi, propanamide,2,2-dimethyl, trimethylacetarnide
Formula Weight 101.15
Percent Purity ≥98%
Chemical Name or Material 2,2,2-Trimethylacetamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.