missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroiodoethane, 99%

Product Code. 11443398
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11443398 50 g 50g
11433398 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11443398 Supplier Thermo Scientific Alfa Aesar Supplier No. A17804.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 353-83-3 | C2H2F3I | 209.938 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 353-83-3
Molecular Formula C2H2F3I
Molecular Weight (g/mol) 209.938
MDL Number MFCD00001082
InChI Key RKOUFQLNMRAACI-UHFFFAOYSA-N
Synonym 2-iodo-1,1,1-trifluoroethane, 2,2,2-trifluoroethyl iodide, trifluoroethyl iodide, ethane, 1,1,1-trifluoro-2-iodo, 1-iodo-2,2,2-trifluoroethane, 2,2,2-trifluoroiodoethane, cf3ch2i, unii-ca0j38we9z, 1,1,1-trifluoroiodoethane, 1,1,1-trifluoro-2-iodo-ethane
PubChem CID 67709
IUPAC Name 1,1,1-trifluoro-2-iodoethane
SMILES C(C(F)(F)F)I

Specifications

Density 2.142
Boiling Point 53°C to 55°C
Refractive Index 1.4009
Quantity 50 g
Beilstein 1733201
Sensitivity Light sensitive
Formula Weight 209.94
Percent Purity 99%
Chemical Name or Material 2,2,2-Trifluoroiodoethane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.