missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,2-Trifluoroethylhydrazine, 65 wt% aqueous solution
Shop All Thermo Scientific Chemicals ProductsDescription
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 65.0, 25.0 |
| CAS Max % | 75.0, 35.0 |
| Color | Colorless to Yellow |
| Density | 1.2940g/mL |
| Flash Point | 42°C |
| Packaging | Glass bottle |
| Linear Formula | CF3CH2NHNH2 |
| Refractive Index | 1.3600 to 1.364 |
| Quantity | 25 g |
| Specific Gravity | 1.294 |
| Show More |
Safety and Handling
Product Identifier
- 2,2,2-Trifluoroethylhydrazine, 70 wt% aqueous solution
- Warning
- Acute toxicity Category 4
- Flammable liquid Category 3
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?