Learn More
2,2,2-Trifluoroethylhydrazine, 70 wt% aqueous solution, Thermo Scientific Chemicals
£23.74 - £266.00
Chemical Identifiers
CAS | 5042-30-8, 7732-18-5 |
---|---|
Molecular Formula | C2H5F3N2 |
Molecular Weight (g/mol) | 114.07 |
MDL Number | MFCD00007622 |
InChI Key | OPMFFAOEPFATTG-UHFFFAOYSA-N |
Synonym | 2,2,2-trifluoroethyl hydrazine, 2,2,2-trifluoroethylhydrazine, 70 % in aqueous solution, 2,2,2-trifluoroethyl hydrazine in water, sntljqhjptadtauup@, acmc-20aoks, trifluoroethyl hydrazine, cf3ch2nhnh2, 2,2,2-trifluorethylhydrazine |
PubChem CID | 78740 |
IUPAC Name | 2,2,2-trifluoroethylhydrazine |
SMILES | C(C(F)(F)F)NN |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10212232
|
Thermo Scientific Acros
365760010 |
1 g | Glass bottle | ||||||
10422985
|
Thermo Scientific Acros
365760050 |
5 g | Glass bottle |
£72.40
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10085153
|
Thermo Scientific Acros
365760250 |
25 g | Glass bottle |
£266.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5042-30-8, 7732-18-5 | |
114.07 | |
OPMFFAOEPFATTG-UHFFFAOYSA-N | |
78740 | |
C(C(F)(F)F)NN |
C2H5F3N2 | |
MFCD00007622 | |
2,2,2-trifluoroethyl hydrazine, 2,2,2-trifluoroethylhydrazine, 70 % in aqueous solution, 2,2,2-trifluoroethyl hydrazine in water, sntljqhjptadtauup@, acmc-20aoks, trifluoroethyl hydrazine, cf3ch2nhnh2, 2,2,2-trifluorethylhydrazine | |
2,2,2-trifluoroethylhydrazine |
Specifications
5042-30-8, 7732-18-5 | |
75.0, 35.0 | |
1.2940g/mL | |
C2H5F3N2 | |
1.3600 to 1.364 | |
1.294 | |
OPMFFAOEPFATTG-UHFFFAOYSA-N | |
2,2,2-trifluoroethylhydrazine | |
78740 | |
≥65% | |
2, 2, 2-Trifluoroethylhydrazine, 70 wt% aqueous solution |
65.0, 25.0 | |
Colorless to Yellow | |
42°C | |
CF3CH2NHNH2 | |
MFCD00007622 | |
2,2,2-trifluoroethyl hydrazine, 2,2,2-trifluoroethylhydrazine, 70 % in aqueous solution, 2,2,2-trifluoroethyl hydrazine in water, sntljqhjptadtauup@, acmc-20aoks, trifluoroethyl hydrazine, cf3ch2nhnh2, 2,2,2-trifluorethylhydrazine | |
C(C(F)(F)F)NN | |
114.07 | |
114.07 | |
Liquid |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Call a POISON CENTER or
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 225-734-
- 2,2,2-Trifluoroethylhydrazine, 70 wt% aqueous solution
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Flammable liquid Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only