missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethylamine hydrochloride, 98%

Product Code. 11464130
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
This item is not returnable. View return policy

Product Code. 11464130

Brand: Thermo Scientific Alfa Aesar A14904.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 373-88-6 | C2H5ClF3N | 135.51 g/mol

2,2,2-Trifluoroethylamine hydrochloride, is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,2-Trifluoroethylamine hydrochloride, is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff

Solubility
It is Soluble in 0.7 to water, but soluble in ethanol and chloroform, slightly soluble in benzene, hardly soluble in ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Hygroscopic. Keep away from strong oxidizing agents, acids, acid chlorides, acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 373-88-6
Molecular Formula C2H5ClF3N
Molecular Weight (g/mol) 135.51
MDL Number MFCD00012875
InChI Key ZTUJDPKOHPKRMO-UHFFFAOYSA-N
Synonym 2,2,2-trifluoroethylamine hydrochloride, 2,2,2-trifluoroethanamine hydrochloride, trifluoroethylamine hydrochloride, ethanamine, 2,2,2-trifluoro-, hydrochloride, 2,2,2-trifluoroethylaminehydrochloride, 2,2,2-trifluoroethylamine, hydrochloride, 2,2,2-trifluoroethan-1-amine hydrochloride, 2,2,2-trifluoroethylamine hcl, ethylamine, trifluoro-, hydrochloride, ethanamine, 2,2,2-trifluoro-, hydrochloride 1:1
PubChem CID 9772
IUPAC Name 2,2,2-trifluoroethanamine;hydrochloride
SMILES [H+].[Cl-].NCC(F)(F)F

Specifications

Melting Point 218°C (sublimation)
Linear Formula CF3CH2NH2·HCl
Quantity 50 g
Beilstein 3652103
Sensitivity Hygroscopic
Solubility Information It is Soluble in 0.7 to water,but soluble in ethanol and chloroform,slightly soluble in benzene,hardly soluble in ether.
Formula Weight 135.52
Percent Purity 98%
Chemical Name or Material 2,2,2-Trifluoroethylamine hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.