missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethyl Tosylate, TRC

Product Code. 30457006
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457006 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457006 Supplier Toronto Research Chemicals Supplier No. TRCT790230250G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,2,2-Trifluoroethyl Tosylate
CAS 433-06-7
Molecular Formula C9H9F3O3S
Quantity 250 g
Synonym 2,2,2-trifluoroethanol 4-Methylbenzenesulfonate, 2,2,2-Trifluoroethanol p-Toluenesulfonate, 2,2,2-Trifluoroethanol 4-Methylbenzenesulfonate, 2,2,2-Trifluoroethyl 4-Methylbenzenesulfonate, 2,2,2-Trifluoroethyl 4-Toluenesulfonate, 2,2,2-Trifluoroethyl p-Toluenesulfonate, NSC 77401, Toluene-4-sulfonic Acid 2,2,2-Trifluoroethyl Ester, p-Toluenesulfonic Acid 2,2,2-Trifluoroethyl Ester
InChI Formula InChI=1S/C9H9F3O3S/c1-7-2-4-8(5-3-7)16(13,14)15-6-9(10,11)12/h2-5H,6H2,1H3
SMILES Cc1ccc(cc1)S(=O)(=O)OCC(F)(F)F
IUPAC Name 2,2,2-trifluoroethyl 4-methylbenzenesulfonate
Molecular Weight (g/mol) 254.23
Formula Weight 254.0224
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.