missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethyl p-toluenesulfonate, 98+%

Product Code. 11493478
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11493478 100 g 100g
11483478 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11493478 Supplier Thermo Scientific Alfa Aesar Supplier No. A17871.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 433-06-7 | C9H9F3O3S | 254.223 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 433-06-7
Molecular Formula C9H9F3O3S
Molecular Weight (g/mol) 254.223
MDL Number MFCD00000443
InChI Key IGKCQDUYZULGBM-UHFFFAOYSA-N
Synonym 2,2,2-trifluoroethyl p-toluenesulfonate, 2,2,2-trifluoroethyl tosylate, trifluoroethanol tosylate, trifluorethyltosylate, 2,2,2-trifluoroethyl-p-toluenesulfonate, ethanol, 2,2,2-trifluoro-, 4-methylbenzenesulfonate, 2,2,2-trifluoroethyl p-toluenesulphonate, ethanol, 2,2,2-trifluoro-, p-toluenesulfonate, 2,2,2-trifluoroethyl 4-methylbenzene-1-sulfonate, ethanol, 2,2,2-trifluoro-, 1-4-methylbenzenesulfonate
PubChem CID 67942
IUPAC Name 2,2,2-trifluoroethyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCC(F)(F)F

Specifications

Melting Point 37°C to 42°C
Boiling Point 87°C to 92°C (0.1 mmHg)
Flash Point >110°C (230°F)
Quantity 100 g
Beilstein 2699394
Sensitivity Moisture sensitive
Formula Weight 254.23
Percent Purity ≥98%
Chemical Name or Material 2,2,2-Trifluoroethyl p-toluenesulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.