missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethyl methyl ether, 99+%

Product Code. 11304815
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11304815 5 g 5g
11314815 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.
Product Code. 11304815 Supplier Thermo Scientific Alfa Aesar Supplier No. L17887.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.

CAS: 460-43-5 | C3H5F3O | 114.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 460-43-5
Molecular Formula C3H5F3O
Molecular Weight (g/mol) 114.07
MDL Number MFCD00236114
InChI Key OHLVGBXMHDWRRX-UHFFFAOYSA-N
Synonym 2,2,2-trifluoroethyl methyl ether, cf3ch2och3, ethane, 1,1,1-trifluoro-2-methoxy, acmc-1csff, trifluoroethyl methyl ether, 2,2,2-trifluoroethyl methyl ethe, 1,1,1-trifluoro-2-methoxy-ethane, #, ethane,1,1,1-trifluoro-2-methoxy, 1,1,1-tris fluoranyl-2-methoxy-ethane
PubChem CID 136304
IUPAC Name 1,1,1-trifluoro-2-methoxyethane
SMILES COCC(F)(F)F

Specifications

Density 1.156
Boiling Point 30°C
Flash Point −8°C (−18°F)
Odor Ether-like
Linear Formula CH3OCH2CF3
Refractive Index 1.2942
Quantity 5 g
UN Number UN1993
Beilstein 1734776
Formula Weight 114.07
Percent Purity ≥99%
Chemical Name or Material 2,2,2-Trifluoroethyl methyl ether
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.