missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethyl methacrylate, 98%, stab. with 30-50 ppm 4-methoxyphenol

Product Code. 15447515
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15447515 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15447515 Supplier Thermo Scientific Alfa Aesar Supplier No. A17012.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 352-87-4 | C6H7F3O2 | 168.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 352-87-4
Density 1.181
Boiling Point 29°C to 30°C (40 mmHg)
Flash Point 25°C (77°F)
Odor Pungent
Molecular Formula C6H7F3O2
Refractive Index 1.361
MDL Number MFCD00013576,MFCD00274739
Quantity 25 g
UN Number UN3272
Beilstein 4175872
Synonym 2,2,2-trifluoroethyl methacrylate, trifluoroethyl methacrylate, methacrylic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethylmethacrylate, 2-propenoic acid, 2-methyl-, 2,2,2-trifluoroethyl ester, 2-methyl-acrylic acid 2,2,2-trifluoro-ethyl ester, 2-propenoic acid, 2-methyl-, trifluoroethyl ester, 2-methyl-2-propenoic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethyl m, acmc-1af8z
InChI Key QTKPMCIBUROOGY-UHFFFAOYSA-N
SMILES CC(=C)C(=O)OCC(F)(F)F
IUPAC Name 2,2,2-trifluoroethyl 2-methylprop-2-enoate
Molecular Weight (g/mol) 168.12
PubChem CID 9608
Formula Weight 168.12
Percent Purity 98%
Chemical Name or Material 2,2,2-Trifluoroethyl methacrylate, stabilized with 30 to 50 ppm 4-methoxyphenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.