missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethyl formate, 95%, Thermo Scientific Chemicals

Product Code. 11332199
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11332199 5 g 5g
11322199 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332199 Supplier Thermo Scientific Alfa Aesar Supplier No. H27212.06
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 32042-38-9 | Molecular Formula: C3H3F3O2 | Molecular Weight: 128.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32042-38-9
Molecular Formula C3H3F3O2
Molecular Weight (g/mol) 128.05
MDL Number MFCD06797733
InChI Key CAFROQYMUICGNO-UHFFFAOYSA-N
Synonym tfef, formic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethylformate, trifluoroethyl formate, zlchem 326, hcooch2cf3, acmc-209hq8, ksc495g7p, 2,2,2-trifluoroethyl formate, 2,2,2-tris fluoranyl ethyl methanoate
PubChem CID 141681
IUPAC Name 2,2,2-trifluoroethyl formate
SMILES FC(F)(F)COC=O

Specifications

Density 1.317
Boiling Point 59°C to 60°C
Flash Point 7°C (45°F)
Refractive Index 1.001
Quantity 5 g
UN Number UN3272
Formula Weight 128.05
Percent Purity 95%
Chemical Name or Material 2,2,2-Trifluoroethyl formate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.