missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethyl acrylate, 98%, stab. with 200ppm 4-methoxyphenol

Product Code. 11423767
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11423767 25 g 25g
11413767 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11423767 Supplier Thermo Scientific Alfa Aesar Supplier No. A18235.14
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 407-47-6 | C5H5F3O2 | 154.088 g/mol

It is used for adhesives because of its peeling property and also used for contact lens to give antifouling property. It is also employed as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used for adhesives because of its peeling property and also used for contact lens to give antifouling property. It is also employed as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents, heat, light UV radiation and free radical initiators. Store under inert gas. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from light and electrostatic charges. Keep it refrigerated.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 407-47-6
Molecular Formula C5H5F3O2
Molecular Weight (g/mol) 154.088
MDL Number MFCD00000444
InChI Key VBHXIMACZBQHPX-UHFFFAOYSA-N
Synonym 2,2,2-trifluoroethyl acrylate, 2-propenoic acid, 2,2,2-trifluoroethyl ester, 2-propenoic acid, trifluoroethyl ester, 2,2,2-trifluoroethylacrylate, acrylic acid, 2,2,2-trifluoroethyl ester, acrylic acid 2,2,2-trifluoroethyl ester, tfol-a, acmc-20aokw, trifluoroethyl acrylate, pubchem12647
PubChem CID 67889
IUPAC Name 2,2,2-trifluoroethyl prop-2-enoate
SMILES C=CC(=O)OCC(F)(F)F

Specifications

Density 1.216
Boiling Point 91°C to 93°C
Flash Point 12°C (53°F)
Odor Pungent
Linear Formula CF3CH2O2CCH=CH2
Refractive Index 1.35
Quantity 25 g
UN Number UN3272
Beilstein 1756611
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 154.09
Percent Purity 98%
Chemical Name or Material 2,2,2-Trifluoroethyl acrylate, stabilized with 200ppm 4-methoxyphenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.