missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethyl acetate, 97%

Product Code. 11363806
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11363806

Brand: Thermo Scientific Alfa Aesar L16871.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 406-95-1 | C4H5F3O2 | 142.08 g/mol

2,2,2-Trifluoroethyl trifluoroacetate is used in the preparation of dimethyl (3,3,3-trifluoro-2,2-dihydroxypropyl)phosphonate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,2-Trifluoroethyl trifluoroacetate is used in the preparation of dimethyl (3,3,3-trifluoro-2,2-dihydroxypropyl)phosphonate

Solubility
Soluble in water (16 g/L) (25°C)

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 406-95-1
Molecular Formula C4H5F3O2
Molecular Weight (g/mol) 142.08
MDL Number MFCD00198018
InChI Key ZOWSJJBOQDKOHI-UHFFFAOYSA-N
Synonym trifluoroethyl acetate, unii-en07rvl78u, en07rvl78u, acetic acid, 2,2,2-trifluoroethyl ester, acetic acid 2,2,2-trifluoroethyl ester, trifluoroethylacetate, 2,2,2-trifluoroethylacetate, acmc-20aokx, cf3ch2o2cch3, ksc494s2j
PubChem CID 136256
IUPAC Name 2,2,2-trifluoroethyl acetate
SMILES CC(=O)OCC(F)(F)F

Specifications

Density 1.258
Boiling Point 78°C
Flash Point 11°C (51°F)
Linear Formula CH3CO2CH2CF3
Refractive Index 1.32
Quantity 25 g
UN Number UN3272
Beilstein 1757502
Solubility Information Soluble in water (16g/L) (25°C)
Formula Weight 142.08
Percent Purity 97%
Chemical Name or Material 2,2,2-Trifluoroethyl acetate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.