missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethanol-d3, for NMR, 99+% atom D, ACROS Organics™

Product Code. 10033440 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10033440 1 g Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10033440 Supplier Thermo Scientific Acros Supplier No. 278780010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Specifications

Physical Form Liquid
Color Colorless
Density 1.4500g/mL
Deuterium (D) 99.0% min.
Packaging Glass Bottle
Quantity 1 g
CAS 77253-67-9
Molecular Formula C2H3F3O
Linear Formula CF3CD2OD
MDL Number MFCD00037682
Synonym 2,2,2-trifluoroethanol-d3, 1,1-2h2-2,2,2-trifluoroetane-1-2h ol, ethan-1,1-d2-ol-d, 2,2,2-trifluoro, trifluoro 2 h? ethan 2 h ol, 2,2,2-trifluoroethanol-d3 >98 atom % d, 2,2,2-trifluoroethanol-d3, >=99.5 atom % d, 2,2,2-trifluoroethanol-d3, 99 atom % d cp
InChI Key RHQDFWAXVIIEBN-IDPMSXFZSA-N
SMILES [2H]OC([2H])([2H])C(F)(F)F
IUPAC Name trifluoro(²H₂)ethan-1-(²H)ol
Molecular Weight (g/mol) 103.06
PubChem CID 2724851
Formula Weight 103.06
Grade NMR
Chemical Name or Material 2, 2, 2-Trifluoroethanol-d3, 99+ atom% D
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.