missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethanol, 99.8%, extra pure, Thermo Scientific Chemicals

Catalog Number p-4509986
19.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
25 mL
100 mL
500 mL
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
75-89-8
C2H3F3O
100.04
MFCD00004672
RHQDFWAXVIIEBN-UHFFFAOYSA-N
trifluoroethanol, ethanol, 2,2,2-trifluoro, 2,2,2-trifluoroethan-1-ol, fluorinol 85, 2,2,2-trifluoroethyl alcohol, trifluoroethyl alcohol, trifluoro ethanol, tfe, perfluoro-1,1-dihydroethanol, tfetoh
6409
CHEBI:42330
2,2,2-trifluoroethanol
OCC(F)(F)F
75-89-8
C2H3F3O
100.04
MFCD00004672
RHQDFWAXVIIEBN-UHFFFAOYSA-N
trifluoroethanol, ethanol, 2,2,2-trifluoro, 2,2,2-trifluoroethan-1-ol, fluorinol 85, 2,2,2-trifluoroethyl alcohol, trifluoroethyl alcohol, trifluoro ethanol, tfe, perfluoro-1,1-dihydroethanol, tfetoh
6409
CHEBI:42330
2,2,2-trifluoroethanol
OCC(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -45.0°C
Color Colorless
Density 1.3900g/mL
pH 5.0 to 7.5 (10% v/v soln.)
Boiling Point 77.0°C to 80.0°C
Flash Point 29°C
Infrared Spectrum Authentic
Assay Percent Range 99.75% min. (GC)
Packaging Glass bottle
Linear Formula CF3CH2OH
Refractive Index 1.2907
Quantity 25 mL
Residue after Evaporation 0.005% max.
Merck Index 15,9848
Specific Gravity 1.39
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohols,ketons,esters and ethers
Viscosity 1.75 mPa.s (25°C)
Formula Weight 100.04
Percent Purity 99.8%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material 2, 2, 2-Trifluoroethanol
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,2,2-Trifluoroethanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H360F-May damage fertility.
  • H373-May cause damage to organs.
  • H301-Toxic if swallowed.
  • H331-Toxic if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'