missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethanesulfonyl chloride, 95%

Product Code. 10331234 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
2.5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2.5g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10331234

missing translation for 'mfr': Thermo Scientific Acros 305350100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 1648-99-3 | C2H2ClF3O2S | 182.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1648-99-3
Molecular Formula C2H2ClF3O2S
Molecular Weight (g/mol) 182.55
MDL Number MFCD00007458
InChI Key CXCHEKCRJQRVNG-UHFFFAOYSA-N
Synonym tresyl chloride, 2,2,2-trifluoroethane-1-sulfonyl chloride, ethanesulfonyl chloride, 2,2,2-trifluoro, 2,2,2-trifluoroethanesulphonyl chloride, 2,2,2-trifluoroethyl sulphonyl chloride, 2,2,2-trifluoroethylsulfonyl chloride, chloro 2,2,2-trifluoroethyl sulfone, 2,2,2-trifluoroethanesulfonylchloride, ethanesulfonylchloride, 2,2,2-trifluoro, pubchem2553
PubChem CID 74242
IUPAC Name 2,2,2-trifluoroethanesulfonyl chloride
SMILES C(C(F)(F)F)S(=O)(=O)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 1.6500g/mL
Boiling Point 64°C to 67°C (45.0 mmHg)
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (ex Cl) (Argentometry)
Packaging Glass bottle
Linear Formula CF3CH2SO2Cl
Refractive Index 1.3870 to 1.389
Quantity 10 g
Specific Gravity 1.65
Formula Weight 182.55
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2, 2, 2-Trifluoroethanesulfonyl chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.