missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroacetamidine, 85%, Thermo Scientific™

Product Code. 15204388 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15204388 1 g 1g
15214388 5 g 5g
15224388 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15204388 Supplier Thermo Scientific Acros Supplier No. 454690010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Kemiske identifikatorer

CAS 354-37-0
Molecular Formula C2H3F3N2
Molecular Weight (g/mol) 112.06
MDL Number MFCD00041545
InChI Key NITMACBPVVUGOJ-UHFFFAOYSA-N
Synonym trifluoroacetamidine, trifluoroethanimidamide, 2,2,2-trifluoroacetamidine, ethanimidamide, 2,2,2-trifluoro, sntljqhjptadtaetp@, trifluoroacetoamidine, trifluoroacetamideine, trifluoro-acetamidine, acmc-20aje4, 2,2,2-trifluoroacetoamidine
PubChem CID 2776882
IUPAC Name 2,2,2-trifluoroethanimidamide
SMILES NC(=N)C(F)(F)F

Tekniske data

Boiling Point 35.0°C to 37.0°C (11.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 84% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 112.05
Percent Purity 85%
Chemical Name or Material 2,2,2-Trifluoroacetamidine
compliance-icons
Produktidentifikator
  • 2,2,2-Trifluoroacetamidine, 85%
Signalord
  • Danger
Farekategori
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Faresætning
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Sikkerhedssætning
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.