missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoro-N-methylacetamide, 98%

Product Code. 11456696
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11456696 25 g 25g
11446696 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11456696 Supplier Thermo Scientific Alfa Aesar Supplier No. B23851.14
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 815-06-5 | C3H4F3NO | 127.066 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 815-06-5
Molecular Formula C3H4F3NO
Molecular Weight (g/mol) 127.066
MDL Number MFCD00009670
InChI Key IQNHBUQSOSYAJU-UHFFFAOYSA-N
Synonym n-methyltrifluoroacetamide, n-methyl-2,2,2-trifluoroacetamide, acetamide, 2,2,2-trifluoro-n-methyl, 2,2,2-trifluoro-n-methyl-acetamide, n-metnyl-2,2,2-trifluoroacetamide, methyltrifluoroacetamide, acmc-209plm, n-methyl-trifluoroacetamide, n-methyltrifluoro-acetamide
PubChem CID 69948
IUPAC Name 2,2,2-trifluoro-N-methylacetamide
SMILES CNC(=O)C(F)(F)F

Specifications

Melting Point 49°C to 52°C
Boiling Point 156°C to 157°C
Flash Point 73°C (163°F)
Linear Formula CF3CONHCH3
Quantity 25 g
Beilstein 1703392
Sensitivity Moisture sensitive
Formula Weight 127.07
Percent Purity ≥98%
Chemical Name or Material 2,2,2-Trifluoro-N-methylacetamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.