missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trichloroethyl Chloroformate, TRC

Product Code. 30453433
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30453433 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30453433 Supplier Toronto Research Chemicals Supplier No. TRCT7743401G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,2,2-Trichloroethyl Chloroformate
CAS 17341-93-4
Molecular Formula C3H2Cl4O2
Quantity 1 g
Synonym Carbonochloridic Acid 2,2,2-Trichloroethyl Ester, Chloro-formic Acid 2,2,2-Trichloroethyl Ester, 2,2,2-Trichloro-ethanol Chloroformate, (Trichloroethoxy)chloroformate, 2,2,2-Trichloroethoxycarbonyl Chloride, Chloroformic Acid 2,2,2-Trichloroethyl Ester, NSC 152038, Troc-Cl, beta,beta,beta-Trichloroethoxycarbonyl Chloride, beta,beta,beta-Trichloroethyl Chloroformate
InChI Formula InChI=1S/C3H2Cl4O2/c4-2(8)9-1-3(5,6)7/h1H2
SMILES ClC(=O)OCC(Cl)(Cl)Cl
IUPAC Name 2,2,2-trichloroethyl carbonochloridate
Molecular Weight (g/mol) 211.86
Formula Weight 209.8809
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.