missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trichloroethyl chloroformate, 97%

Product Code. 10257743 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10257743 25 g Glass bottle 25g
10522761 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10257743 Supplier Thermo Scientific Acros Supplier No. 161810250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17341-93-4 | C3H2Cl4O2 | 211.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17341-93-4
MDL Number MFCD00000810
InChI Key LJCZNYWLQZZIOS-UHFFFAOYSA-N
Synonym 2,2,2-trichloroethyl chloroformate, trichloroethyl chloroformate, 2,2,2-trichloroethylchloroformate, carbonochloridic acid, 2,2,2-trichloroethyl ester, 2,2,2-trichloroethoxycarbonyl chloride, chloroformic acid 2,2,2-trichloroethyl ester, formic acid, chloro-, 2,2,2-trichloroethyl ester, chloro 2,2,2-trichloroethoxy methanone, beta, beta, beta-trichloroethoxycarbonyl chloride, troc
PubChem CID 87063
IUPAC Name 2,2,2-trichloroethyl carbonochloridate
SMILES C(C(Cl)(Cl)Cl)OC(=O)Cl

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 1°C
Color Yellow
Density 1.5390g/mL
Boiling Point 171°C to 172°C
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C3H2Cl4O2
Linear Formula ClCO2CH2CCl3
Refractive Index 1.4690 to 1.471
Quantity 25 g
Fieser 02,426; 03,296; 04,522; 05,686; 07,383
Specific Gravity 1.539
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in most common organic solvents
Molecular Weight (g/mol) 211.86
Formula Weight 211.86
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2, 2, 2-Trichloroethyl chloroformate, 96%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,2,2-Trichloroethyl chloroformate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P402+P404-Store in a dry place. Store in a closed container.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.