missing translation for 'onlineSavingsMsg'
Learn More

2-[(2,2-Dimethylpropanoyl)amino]nicotinic acid, 97%, Thermo Scientific™

Product Code. 10523242
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10523242 1 g 1g
10623413 250 mg 250mg
10718635 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10523242 Supplier Thermo Scientific Maybridge Supplier No. MO00352DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 125867-25-6
Molecular Formula C11H14N2O3
Molecular Weight (g/mol) 222.244
MDL Number MFCD03659710
InChI Key KXLOGMIKCNUSNG-UHFFFAOYSA-N
Synonym 2-pivalamidonicotinic acid, 2-2,2-dimethyl-propionylamino-nicotinic acid, 2-2,2,2-trimethylacetamido nicotinic acid, 2-2,2-dimethylpropanoyl amino nicotinic acid, 2-2,2-dimethylpropanamido pyridine-3-carboxylic acid, 2-pivaloylamino nicotinic acid, 2-2,2-dimethylpropanoylamino pyridine-3-carboxylic acid, maybridge3_004279, acmc-20aorg, pubchem16551
PubChem CID 2779758
IUPAC Name 2-(2,2-dimethylpropanoylamino)pyridine-3-carboxylic acid
SMILES CC(C)(C)C(=O)NC1=C(C=CC=N1)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 222.24
Percent Purity 97%
Chemical Name or Material 2-[(2, 2-Dimethylpropanoyl)amino]nicotinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.