missing translation for 'onlineSavingsMsg'
Learn More

2-(2,2-Difluorobenzo[d][1,3]dioxol-5-yl)acetonitrile, TRC

Product Code. 30383898
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383898 50 mg -
30386014 500 mg 500mg
30378491 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30383898 Supplier Toronto Research Chemicals Supplier No. TRCD45209350MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68119-31-3
Molecular Formula C9H5NO2F2
Molecular Weight (g/mol) 197.14
SMILES FC1(F)Oc2ccc(CC#N)cc2O1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C9H5F2NO2/c10-9(11)13-7-2-1-6(3-4-12)5-8(7)14-9/h1-2,5H,3H2
Formula Weight 197.0288
Chemical Name or Material 2-(2,2-Difluorobenzo[d][1,3]dioxol-5-yl)acetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.