missing translation for 'onlineSavingsMsg'
Learn More

2-(2,2,4,7-Tetramethyl-1,2,3,4-tetrahydroquinolin-1-yl)ethan-1-ol, 97%, Thermo Scientific™

Product Code. 11351416
Change view
Click to view available options
Quantity:
25 g
50 g
Packaging:
Amber Glass Bottle
Unit Size:
25g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11351416 50 g Amber Glass Bottle 50g
11370547 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11351416 Supplier Thermo Scientific Maybridge Supplier No. BTB13839EE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 53817-44-0
Molecular Formula C15H23NO
Molecular Weight (g/mol) 233.355
InChI Key DICAWUKDDRKZDZ-UHFFFAOYSA-N
Synonym 2-2,2,4,7-tetramethyl-1,2,3,4-tetrahydroquinolin-1-yl ethan-1-ol, 1 2h-quinolineethanol, 3,4-dihydro-2,2,4,7-tetramethyl, 1-2-hydroxyethyl-1,2,3,4-tetrahydro-2,2,4,7-tetramethylquinoline, 2-2,2,4,7-tetramethyl-3,4-dihydroquinolin-1-yl ethanol, acmc-1amas, maybridge1_002615, 3,4-dihydro-2,2,4,7-tetramethyl-2h-quinoline-1-ethanol, 1 2h-quinolineethanol,3,4-dihydro-2,2,4,7-tetramethyl, 3,4-dihydro-2,2,4,7-tetramethyl-1 2h-quinolineethanol, 1,2,3,4-tetrahydro-2,2,4,7-tetramethyl-1-quinolineethanol
PubChem CID 103845
IUPAC Name 2-(2,2,4,7-tetramethyl-3,4-dihydroquinolin-1-yl)ethanol
SMILES CC1CC(N(C2=C1C=CC(=C2)C)CCO)(C)C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 50 g
Formula Weight 233.35
Percent Purity 97%
Chemical Name or Material 2-(2,2,4,7-Tetramethyl-1,2,3,4-tetrahydroquinolin-1-yl)ethan-1-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.